ENA311760780 Enamine

6-chloro-2-oxo-1,2-dihydroquinoline-4-carboxylic acid

  • MDL number MFCD18089498
  • Cas Number : 32431-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 315-316
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72404
Availability details for
Quantity
  • Availability
  • Add to Cart