ENA311760781 Enamine

3-(1H-imidazol-4-yl)propan-1-ol hydrochloride

  • MDL number MFCD09908716
  • Cas Number : 152028-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71576

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71576
Availability details for
Quantity
  • Availability
  • Add to Cart