ENA311760782 Enamine

1-{2-[(5-chloropyridin-2-yl)(methyl)amino]acetyl}piperidine-3-carboxylic acid

  • MDL number MFCD13668536
  • Cas Number : 1099164-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72406
Availability details for
Quantity
  • Availability
  • Add to Cart