ENA311760785 Enamine

6-[(propan-2-yl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD11131987
  • Cas Number : 960060-85-9
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71577
Availability details for
Quantity
  • Availability
  • Add to Cart