ENA311760788 Enamine

4-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)benzoic acid

  • MDL number MFCD18089501
  • Cas Number : 189365-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72414
Availability details for
Quantity
  • Availability
  • Add to Cart