ENA311760792 Enamine

2-bromo-1-(2,4-dimethoxyphenyl)propan-1-one

  • MDL number MFCD11182357
  • Cas Number : 117844-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72432
Availability details for
Quantity
  • Availability
  • Add to Cart