ENA311760798 Enamine

2-phenyl-1,3-oxazole-4-carbaldehyde

  • MDL number MFCD06738551
  • Cas Number : 20771-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72448

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72448
Availability details for
Quantity
  • Availability
  • Add to Cart