ENA311760799 Enamine

2-fluoro-6-(1H-1,2,4-triazol-1-yl)aniline

  • MDL number MFCD12149615
  • Cas Number : 1183856-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72451

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72451
Availability details for
Quantity
  • Availability
  • Add to Cart