ENA311760803 Enamine

[3-fluoro-4-(methoxymethyl)phenyl]methanamine

  • MDL number MFCD09816938
  • Cas Number : 1016685-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71705
Availability details for
Quantity
  • Availability
  • Add to Cart