ENA311760809 Enamine

2-{1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-oxo-4,5-dihydro-1H-pyrazol-3-yl}acetic acid

  • MDL number MFCD13665465
  • Cas Number : 1097785-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72463
Availability details for
Quantity
  • Availability
  • Add to Cart