ENA311760811 Enamine

2-(methylamino)-1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD13562036
  • Cas Number : 1220036-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72465
Availability details for
Quantity
  • Availability
  • Add to Cart