ENA311760812 Enamine

1-(5-chloro-2,3-dihydro-1-benzofuran-2-carbonyl)piperidine-3-carboxylic acid

  • MDL number MFCD13668569
  • Cas Number : 1098369-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72466
Availability details for
Quantity
  • Availability
  • Add to Cart