ENA311760814 Enamine

2-(2-chloro-6-fluorobenzamido)-5-methylbenzoic acid

  • MDL number MFCD13373379
  • Cas Number : 1040341-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72469
Availability details for
Quantity
  • Availability
  • Add to Cart