ENA311760815 Enamine

2-[(4-formylphenoxy)methyl]furan-3-carboxylic acid

  • MDL number MFCD12072646
  • Cas Number : 1155071-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72470
Availability details for
Quantity
  • Availability
  • Add to Cart