ENA311760820 Enamine

2-(3-formylphenoxy)-2-phenylacetic acid

  • MDL number MFCD12754249
  • Cas Number : 1094672-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72476
Availability details for
Quantity
  • Availability
  • Add to Cart