ENA311760821 Enamine

3-{[(tert-butoxy)carbonyl]amino}-4-fluorobenzoic acid

  • MDL number MFCD12730844
  • Cas Number : 1184472-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71720

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71720
Availability details for
Quantity
  • Availability
  • Add to Cart