ENA311760826 Enamine

1-(azetidin-1-yl)prop-2-en-1-one

  • MDL number MFCD12091259
  • Cas Number : 98501-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72484

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72484
Availability details for
Quantity
  • Availability
  • Add to Cart