ENA311760827 Enamine

1-(4-methoxyphenyl)-2,2-dimethylpropan-1-one

  • MDL number MFCD03841166
  • Cas Number : 2040-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72485
Availability details for
Quantity
  • Availability
  • Add to Cart