ENA311760830 Enamine

1-[(benzyloxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD00991046
  • Cas Number : 91958-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72490
Availability details for
Quantity
  • Availability
  • Add to Cart