ENA311760838 Enamine

2-(4-formyl-2-methoxy-5-nitrophenoxy)acetic acid

  • MDL number MFCD11536175
  • Cas Number : 90429-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71738
Availability details for
Quantity
  • Availability
  • Add to Cart