ENA311760842 Enamine

(1S)-1-(4-iodophenyl)ethan-1-ol

  • MDL number MFCD09863731
  • Cas Number : 104013-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72506

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72506
Availability details for
Quantity
  • Availability
  • Add to Cart