ENA311760844 Enamine

2-(piperidin-2-yl)-1H-indole

  • MDL number MFCD02663628
  • Cas Number : 20862-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-135
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72509
Availability details for
Quantity
  • Availability
  • Add to Cart