ENA311760845 Enamine

2-phenoxy-1-phenylethan-1-amine hydrochloride

  • MDL number MFCD17214612
  • Cas Number : 16797-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72510
Availability details for
Quantity
  • Availability
  • Add to Cart