ENA311760853 Enamine

2-(morpholin-4-yl)ethanethioamide

  • MDL number MFCD09948450
  • Cas Number : 16888-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71742

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71742
Availability details for
Quantity
  • Availability
  • Add to Cart