ENA311760854 Enamine

2-(4-aminopiperidin-1-yl)acetonitrile

  • MDL number MFCD12137384
  • Cas Number : 1154259-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71743
Availability details for
Quantity
  • Availability
  • Add to Cart