ENA311760856 Enamine

4-(morpholin-4-yl)butan-2-ol

  • MDL number MFCD16100730
  • Cas Number : 858440-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72530

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72530
Availability details for
Quantity
  • Availability
  • Add to Cart