ENA311760858 Enamine

[(4-bromophenyl)(pyridin-3-yl)methyl](ethyl)amine

  • MDL number MFCD12419528
  • Cas Number : 1153742-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72532
Availability details for
Quantity
  • Availability
  • Add to Cart