ENA311760868 Enamine

6-(3,4-dimethoxyphenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD12210507
  • Cas Number : 78558-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72544
Availability details for
Quantity
  • Availability
  • Add to Cart