ENA311760870 Enamine

2-(2-bromo-4-chlorophenoxy)-2-phenylacetic acid

  • MDL number MFCD09717589
  • Cas Number : 938259-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71747
Availability details for
Quantity
  • Availability
  • Add to Cart