ENA311760871 Enamine

2-fluoro-6-phenoxyaniline

  • MDL number MFCD12149632
  • Cas Number : 1178028-54-0
  • Molecular Weight : 203.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71748
Availability details for
Quantity
  • Availability
  • Add to Cart