ENA311760879 Enamine

ethyl 1-(2-aminophenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD12417499
  • Cas Number : 1155927-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72574
Availability details for
Quantity
  • Availability
  • Add to Cart