ENA311760880 Enamine

1-(4-methylphenyl)-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid

  • MDL number MFCD09048804
  • Cas Number : 223444-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72575
Availability details for
Quantity
  • Availability
  • Add to Cart