ENA311760882 Enamine

{1-[(3,4-dimethoxybenzene)sulfonyl]piperidin-4-yl}methanamine hydrochloride

  • MDL number MFCD18089528
  • Cas Number : 1216563-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72578

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72578
Availability details for
Quantity
  • Availability
  • Add to Cart