ENA311760884 Enamine

4-[(3-hydroxypropyl)sulfanyl]benzoic acid

  • MDL number MFCD11647930
  • Cas Number : 254910-61-7
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72580
Availability details for
Quantity
  • Availability
  • Add to Cart