ENA311760889 Enamine

2-(furan-3-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD18089531
  • Cas Number : 955400-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72592
Availability details for
Quantity
  • Availability
  • Add to Cart