ENA311760892 Enamine

(4-chlorophenyl)(pyridin-3-yl)methanol

  • MDL number MFCD03076148
  • Cas Number : 68885-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71757
Availability details for
Quantity
  • Availability
  • Add to Cart