ENA311760894 Enamine

3-(prop-2-yn-1-yloxy)phenol

  • MDL number MFCD11103469
  • Cas Number : 3692-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71758
Availability details for
Quantity
  • Availability
  • Add to Cart