ENA311760903 Enamine

4,4-dimethyl-1-(4-methylphenyl)pentane-1,3-dione

  • MDL number MFCD11185175
  • Cas Number : 41070-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71760
Availability details for
Quantity
  • Availability
  • Add to Cart