ENA311760905 Enamine

methyl 3-methyl-4-[(propan-2-yl)amino]benzoate

  • MDL number MFCD12041277
  • Cas Number : 1157956-62-1
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71763
Availability details for
Quantity
  • Availability
  • Add to Cart