ENA311760913 Enamine

2-(thiophen-2-ylmethoxy)benzoic acid

  • MDL number MFCD09814271
  • Cas Number : 67443-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71822

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71822
Availability details for
Quantity
  • Availability
  • Add to Cart