ENA311760918 Enamine

2-(4-{[(9H-fluoren-9-yl)methoxy]carbonyl}morpholin-3-yl)acetic acid

  • MDL number MFCD04114893
  • Cas Number : 885273-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71827
Availability details for
Quantity
  • Availability
  • Add to Cart