ENA311760920 Enamine

2-[2-(benzyloxy)phenoxy]acetic acid

  • MDL number MFCD00598600
  • Cas Number : 244123-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71830

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71830
Availability details for
Quantity
  • Availability
  • Add to Cart