ENA311760921 Enamine

thiophene-3-carboximidamide hydrochloride

  • MDL number MFCD01940341
  • Cas Number : 51528-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71831

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71831
Availability details for
Quantity
  • Availability
  • Add to Cart