ENA311760935 Enamine

2-(4-bromo-2-chlorophenoxy)-2-phenylacetic acid

  • MDL number MFCD09711097
  • Cas Number : 938261-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71839
Availability details for
Quantity
  • Availability
  • Add to Cart