ENA311760942 Enamine

3-(2,4-dichlorophenyl)-2-methylpropanoic acid

  • MDL number MFCD12794490
  • Cas Number : 119411-79-9
  • Molecular Weight : 233.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71841
Availability details for
Quantity
  • Availability
  • Add to Cart