ENA311760947 Enamine

1-(2-fluoro-4-methylphenyl)-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD12559985
  • Cas Number : 1039900-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71846
Availability details for
Quantity
  • Availability
  • Add to Cart