ENA311760951 Enamine

N-(5-chloro-2-hydroxyphenyl)-4-methylbenzene-1-sulfonamide

  • MDL number MFCD02173764
  • Cas Number : 136061-90-0
  • Molecular Weight : 297.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71850
Availability details for
Quantity
  • Availability
  • Add to Cart