ENA311760957 Enamine

2-bromo-1,3-benzoxazole

  • MDL number MFCD12780021
  • Cas Number : 68005-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71857

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71857
Availability details for
Quantity
  • Availability
  • Add to Cart