ENA311760960 Enamine

3-(5-bromothiophen-2-yl)-3-oxopropanenitrile

  • MDL number MFCD07785836
  • Cas Number : 71683-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71591
Availability details for
Quantity
  • Availability
  • Add to Cart