ENA311760965 Enamine

N-ethylsulfamoyl chloride

  • MDL number MFCD12401754
  • Cas Number : 16548-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71869
Availability details for
Quantity
  • Availability
  • Add to Cart